Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc25a27
Entrez Gene ID
85262
Gene Name
solute carrier family 25, member 27
Gene Aliases
Ucp4
Chromosome Location
Chr.9: 19880469 - 19907506 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053500.1 NP_445952.1 1 - 2 156 95
XM_006244610.2 XP_006244672.1 1 - 2 272 95
XM_006244611.2 XP_006244673.1 1 - 2 272 95
XM_006244612.2 XP_006244674.1 1 - 2 272 95
GenBank mRNA AJ300162.1 - 1 - 2 156 95
AJ300163.1 - 1 - 2 243 95
AJ300164.1 - 1 - 2 264 95

View Full Product Details