The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053541.1 | NP_445993.1 | 2 - 3 | 490 | 66 |
XM_008759200.1 | XP_008757422.1 | 2 - 3 | 569 | 66 | |
XM_008759202.1 | XP_008757424.1 | 2 - 3 | 569 | 66 | |
GenBank mRNA | AB009463.1 | - | 2 - 3 | 490 | 66 |