Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pgf
Entrez Gene ID
94203
Gene Name
placental growth factor
Gene Aliases
Plgf
Chromosome Location
Chr.6: 108994016 - 109004598 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053595.2 NP_446047.1 2 - 3 406 60
GenBank mRNA BC087006.1 - 2 - 3 406 60
L40030.1 - 2 - 3 141 60

View Full Product Details