The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053675.2 | NP_446127.3 | 2 - 3 | 636 | 95 |
XM_006235636.2 | XP_006235698.1 | 2 - 3 | 547 | 95 | |
GenBank mRNA | AF123651.1 | - | 1 - 2 | 437 | 95 |
BC081752.1 | - | 2 - 3 | 636 | 95 |