The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053726.2 | NP_446178.2 | 9 - 10 | 1334 | 96 |
GenBank mRNA | AB126813.1 | - | 9 - 10 | 1334 | 96 |
BC127467.1 | - | 9 - 10 | 1305 | 96 |