The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034117.1 | NP_001029289.1 | 8 - 9 | 1030 | 81 |
NM_053739.2 | NP_446191.1 | 8 - 9 | 1052 | 81 | |
GenBank mRNA | AY033824.1 | - | 7 - 8 | 828 | 81 |
BC074011.1 | - | 8 - 9 | 917 | 81 |