Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkia
Entrez Gene ID
114906
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor alpha
Gene Aliases
-
Chromosome Location
Chr.2: 96593185 - 96668222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053772.2 NP_446224.1 4 - 5 531 94
XM_006232153.2 XP_006232215.1 2 - 3 727 94
XM_006232155.2 XP_006232217.1 3 - 4 378 94
XM_006232156.2 XP_006232218.1 3 - 4 2430 94
XM_008760857.1 XP_008759079.1 3 - 4 855 94
GenBank mRNA FQ211701.1 - 3 - 4 438 94
L02615.1 - 2 - 3 214 94

View Full Product Details