The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053926.2 | NP_446378.1 | 8 - 9 | 1234 | 59 |
GenBank mRNA | AB032899.1 | - | 8 - 9 | 1234 | 59 |