The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053946.1 | NP_446398.1 | 2 - 3 | 318 | 139 |
GenBank mRNA | AF008554.2 | - | 2 - 3 | 318 | 139 |