The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053995.3 | NP_446447.2 | 5 - 6 | 617 | 60 |
XM_006248478.2 | XP_006248540.1 | 5 - 6 | 673 | 60 | |
XM_008768691.1 | XP_008766913.1 | 4 - 5 | 455 | 60 | |
GenBank mRNA | BC085916.1 | - | 5 - 6 | 617 | 60 |
FQ211457.1 | - | 2 - 3 | 171 | 60 |