The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054000.2 | NP_446452.2 | 1 - 2 | 235 | 120 |
GenBank mRNA | M77482.1 | - | 1 - 2 | 238 | 120 |