The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057193.2 | NP_476541.2 | 6 - 7 | 865 | 89 |
XM_006242864.2 | XP_006242926.1 | 6 - 7 | 1043 | 89 | |
GenBank mRNA | AJ305049.1 | - | 6 - 7 | 865 | 89 |
FQ223464.1 | - | 6 - 7 | 886 | 89 |