The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057210.2 | NP_476558.2 | 2 - 3 | 855 | 84 |
XM_006232952.2 | XP_006233014.1 | 3 - 4 | 1284 | 84 | |
GenBank mRNA | BC092132.1 | - | 2 - 3 | 855 | 84 |
L05435.1 | - | 3 - 4 | 1201 | 84 |