Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpr83
Entrez Gene ID
140595
Gene Name
G protein-coupled receptor 83
Gene Aliases
Gir
Chromosome Location
Chr.8: 13365583 - 13376071 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080411.1 NP_536336.1 1 - 2 691 79
GenBank mRNA AY029071.1 - 1 - 2 691 79

View Full Product Details