The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080775.1 | NP_542953.1 | 7 - 8 | 689 | 77 |
XM_008762380.1 | XP_008760602.1 | 7 - 8 | 801 | 77 | |
GenBank mRNA | M83210.1 | - | 7 - 8 | 689 | 77 |