The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080780.2 | NP_542958.2 | 10 - 11 | 1230 | 78 |
XM_006246566.2 | XP_006246628.1 | 9 - 10 | 1238 | 78 | |
GenBank mRNA | X92069.1 | - | 10 - 11 | 1230 | 78 |
X97328.1 | - | 10 - 11 | 1096 | 78 |