The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080898.2 | NP_543174.1 | 5 - 6 | 1703 | 74 |
GenBank mRNA | AY064411.1 | - | - | 2034 | 74 |
EF672039.1 | - | 5 - 6 | 1703 | 74 |