The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133402.2 | NP_596893.1 | 1 - 1 | 620 | 58 |
GenBank mRNA | AB067678.1 | - | 1 - 1 | 550 | 58 |
BC101932.1 | - | 1 - 1 | 620 | 58 |