The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133428.2 | NP_596919.1 | 4 - 5 | 606 | 83 |
GenBank mRNA | AB055895.1 | - | 4 - 5 | 606 | 83 |
BC089779.1 | - | 4 - 5 | 595 | 83 | |
FQ210661.1 | - | 4 - 5 | 606 | 83 | |
FQ219046.1 | - | 2 - 3 | 210 | 83 |