The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033924.1 | NP_001029096.1 | 9 - 10 | 654 | 80 |
NM_133550.2 | NP_598234.2 | 8 - 9 | 591 | 80 | |
XM_008768939.1 | XP_008767161.1 | 8 - 9 | 818 | 80 | |
GenBank mRNA | AF381978.1 | - | 8 - 9 | 540 | 80 |
BC078749.1 | - | 9 - 10 | 654 | 80 | |
X73579.1 | - | 7 - 8 | 474 | 80 |