The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133570.5 | NP_598254.2 | 1 - 2 | 249 | 75 |
GenBank mRNA | BF567195.1 | - | 1 - 2 | 230 | 75 |
M31176.1 | - | 1 - 2 | 653 | 75 |