Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb22
Entrez Gene ID
171412
Gene Name
defensin beta 22
Gene Aliases
2D6glycoprotein
Chromosome Location
Chr.3: 147961818 - 147966097 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134391.1 NP_599218.1 1 - 2 312 91
GenBank mRNA AF329091.1 - 1 - 2 72 91
AJ309150.1 - 1 - 2 312 91
AY621353.1 - 1 - 2 312 91

View Full Product Details