The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134455.1 | NP_604450.1 | 1 - 2 | 90 | 74 |
XM_008772329.1 | XP_008770551.1 | 2 - 3 | 827 | 74 | |
GenBank mRNA | AF030358.2 | - | 1 - 2 | 90 | 74 |
BC070938.1 | - | 1 - 2 | 91 | 74 |