The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134459.1 | NP_604454.1 | 3 - 4 | 257 | 87 |
XM_006251676.2 | XP_006251738.1 | 3 - 4 | 293 | 87 | |
XM_008770596.1 | XP_008768818.1 | 3 - 4 | 283 | 87 | |
GenBank mRNA | AB031014.1 | - | 3 - 4 | 257 | 87 |
AF481858.1 | - | 3 - 4 | 204 | 87 |