The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139085.1 | NP_620785.1 | 2 - 3 | 343 | 96 |
GenBank mRNA | AF501290.1 | - | 2 - 3 | 343 | 96 |