The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139255.3 | NP_640348.2 | 9 - 10 | 1282 | 70 |
GenBank mRNA | AY095481.1 | - | 3 - 4 | 314 | 70 |
BC086408.1 | - | 4 - 5 | 415 | 70 | |
BC097934.1 | - | 5 - 6 | 588 | 70 | |
BC104715.1 | - | 4 - 5 | 353 | 70 | |
BC126078.1 | - | 5 - 6 | 486 | 70 | |
BC161835.1 | - | 5 - 6 | 444 | 70 |