The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139327.1 | NP_647543.1 | 1 - 2 | 139 | 99 |
XM_006256156.1 | XP_006256218.1 | 2 - 3 | 286 | 99 | |
XM_006256157.2 | XP_006256219.1 | 3 - 4 | 399 | 99 | |
XM_006256158.2 | XP_006256220.1 | 2 - 3 | 328 | 99 | |
XM_006256164.1 | XP_006256226.1 | 2 - 3 | 295 | 99 | |
GenBank mRNA | AF507966.1 | - | 1 - 2 | 139 | 99 |