Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc7a6os
Entrez Gene ID
246187
Gene Name
solute carrier family 7, member 6 opposite strand
Gene Aliases
-
Chromosome Location
Chr.19: 38180859 - 38189523 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139328.2 NP_647544.2 3 - 4 681 103
GenBank mRNA AY098917.2 - 2 - 3 349 103
FQ211988.1 - 3 - 4 688 103
FQ220945.1 - 3 - 4 692 103
FQ224990.1 - 2 - 3 413 103
FQ233662.1 - 3 - 4 690 103

View Full Product Details