The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145089.4 | NP_659557.1 | 6 - 7 | 834 | 59 |
GenBank mRNA | AF329099.1 | - | 6 - 7 | 834 | 59 |
AJ427914.2 | - | - | 848 | 59 | |
AJ427915.1 | - | 3 - 4 | 395 | 59 | |
FQ212762.1 | - | 6 - 7 | 808 | 59 |