Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp4x1
Entrez Gene ID
246767
Gene Name
cytochrome P450, family 4, subfamily x, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.5: 133929532 - 133959447 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145675.1 NP_663708.1 9 - 10 1200 69
GenBank mRNA AF439343.1 - 9 - 10 1200 69

View Full Product Details