The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147138.2 | NP_671479.1 | 1 - 2 | 125 | 68 |
GenBank mRNA | AF439397.1 | - | 1 - 2 | 107 | 68 |
AF532776.1 | - | 1 - 2 | 125 | 68 | |
BC063144.1 | - | 1 - 2 | 117 | 68 | |
EU000468.1 | - | 1 - 2 | 93 | 68 |