The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147211.1 | NP_671744.1 | 6 - 7 | 1449 | 62 |
GenBank mRNA | U25281.1 | - | 6 - 7 | 1449 | 62 |