The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152937.2 | NP_690920.1 | 1 - 2 | 328 | 98 |
GenBank mRNA | AF406779.1 | - | 1 - 2 | 328 | 98 |
AJ441127.1 | - | - | 353 | 98 |