The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153306.1 | NP_695218.1 | 1 - 2 | 345 | 58 |
GenBank mRNA | AB036990.1 | - | 1 - 2 | 345 | 58 |