Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc42
Entrez Gene ID
64465
Gene Name
cell division cycle 42
Gene Aliases
-
Chromosome Location
Chr.5: 155690267 - 155728384 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008764287.1 XP_008762509.1 5 - 6 638 95
GenBank mRNA AF205635.1 - 4 - 5 485 95
FQ213007.1 - 5 - 6 557 95

View Full Product Details