Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fxyd3
Entrez Gene ID
116831
Gene Name
FXYD domain-containing ion transport regulator 3
Gene Aliases
MAT8
Chromosome Location
Chr.1: 89502561 - 89509449 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172317.2 NP_758528.1 2 - 3 68 64
XM_006228818.2 XP_006228880.1 2 - 3 318 64
XM_006228819.2 XP_006228881.1 2 - 3 172 64
XM_006228820.2 XP_006228882.1 2 - 3 308 64
GenBank mRNA AW142066.1 - 1 - 2 73 64
CA505583.1 - 6 - 7 64
DY471861.1 - 2 - 3 440 64

View Full Product Details