Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Has3
Entrez Gene ID
266805
Gene Name
hyaluronan synthase 3
Gene Aliases
-
Chromosome Location
Chr.19: 39063319 - 39077745 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172319.1 NP_758822.1 1 - 2 177 77
GenBank mRNA AB097569.1 - 1 - 2 177 77

View Full Product Details