The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173097.1 | NP_775120.1 | 2 - 3 | 104 | 69 |
XM_008763019.1 | XP_008761241.1 | 2 - 3 | 242 | 69 | |
GenBank mRNA | L20869.1 | - | 2 - 3 | 104 | 69 |