The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173145.1 | NP_775168.1 | 2 - 3 | 1200 | 97 |
GenBank mRNA | U67140.1 | - | 2 - 3 | 1200 | 97 |