Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Disc1
Entrez Gene ID
307940
Gene Name
disrupted in schizophrenia 1
Gene Aliases
-
Chromosome Location
Chr.19: 57818865 - 58069992 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175596.2 NP_783186.2 10 - 11 2014 73
XM_008772616.1 XP_008770838.1 10 - 11 3409 73
GenBank mRNA AY177674.2 - 10 - 11 2014 73

View Full Product Details