Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Taar8a
Entrez Gene ID
319104
Gene Name
trace amine-associated receptor 8a
Gene Aliases
Ta11, Trar5
Chromosome Location
Chr.1: 22402878 - 22404002 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175599.1 NP_783189.1 1 - 1 457 106

Gene Symbol
LOC100910174
Entrez Gene ID
100910174
Gene Name
trace amine-associated receptor 8a-like
Gene Aliases
-
Chromosome Location
Chr.1: 22364559 - 22365675 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003748672.3 XP_003748720.1 1 - 1 449 106

Gene Symbol
Taar8b
Entrez Gene ID
319106
Gene Name
trace amine-associated receptor 8b
Gene Aliases
Ta7
Chromosome Location
Chr.1: 23565551 - 23568822 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175601.1 NP_783191.1 1 - 1 367 106
XM_006227718.2 XP_006227780.2 2 - 2 433 106
XM_006227719.2 XP_006227781.1 2 - 2 855 106
XM_008758615.1 XP_008756837.1 3 - 3 1013 106

Gene Symbol
Taar8c
Entrez Gene ID
319105
Gene Name
trace amine-associated receptor 8c
Gene Aliases
Ta10
Chromosome Location
Chr.1: 22332001 - 22350442 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175600.1 NP_783190.1 1 - 1 106
XM_008758669.1 XP_008756891.1 1 - 1 457 106

View Full Product Details