Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbnl2
Entrez Gene ID
680445
Gene Name
muscleblind-like splicing regulator 2
Gene Aliases
-
Chromosome Location
Chr.15: 105640097 - 105797933 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111064.1 NP_001104534.1 4 - 5 1216 79
GenBank mRNA AA818999.1 - 1 - 2 79
BE111982.1 - 1 - 2 79

View Full Product Details