Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnase2b
Entrez Gene ID
59296
Gene Name
deoxyribonuclease II beta
Gene Aliases
Dlad, UOX
Chromosome Location
Chr.2: 252436363 - 252454715 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021664.1 NP_067696.1 2 - 3 448 71
XM_006233464.2 XP_006233526.1 3 - 4 525 71
XM_006233465.2 XP_006233527.1 3 - 4 552 71
XM_006233466.2 XP_006233528.1 3 - 4 555 71
XM_006233468.2 XP_006233530.1 3 - 4 936 71
XM_008761521.1 XP_008759743.1 3 - 4 658 71
GenBank mRNA AF178974.1 - 2 - 3 448 71

View Full Product Details