Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc27a5
Entrez Gene ID
79111
Gene Name
solute carrier family 27 (fatty acid transporter), member 5
Gene Aliases
BAL, rBAL-1
Chromosome Location
Chr.1: 65576599 - 65587192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024143.2 NP_077057.1 3 - 4 1109 72
GenBank mRNA AF242189.1 - 3 - 4 1110 72
BC091147.1 - 3 - 4 1132 72

View Full Product Details