The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031034.2 | NP_112296.1 | 1 - 2 | 337 | 61 |
GenBank mRNA | D85760.1 | - | 1 - 2 | 371 | 61 |
DQ120479.1 | - | 1 - 2 | 306 | 61 | |
DQ120480.1 | - | 1 - 2 | 306 | 61 |