Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm7
Entrez Gene ID
81672
Gene Name
glutamate receptor, metabotropic 7
Gene Aliases
-
Chromosome Location
Chr.4: 142452616 - 143368460 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031040.1 NP_112302.1 3 - 4 1276 80
XM_008763199.1 XP_008761421.1 3 - 4 1276 80
GenBank mRNA D16817.1 - 3 - 4 1276 80
U06832.1 - 3 - 4 1011 80

View Full Product Details