Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Thra
- Entrez Gene ID
-
81812
- Gene Name
- thyroid hormone receptor alpha
- Gene Aliases
-
ERBA1, Thra1, c-erbA-1
- Chromosome Location
-
Chr.10: 86657285 - 86684935 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M31174.1 |
- |
- |
212 |
99 |
View Full Product Details