Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a2
Entrez Gene ID
29503
Gene Name
solute carrier family 22 (organic cation transporter), member 2
Gene Aliases
OCT2, OCT2r, rOCT2
Chromosome Location
Chr.1: 48318025 - 48360219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031584.2 NP_113772.1 6 - 7 1152 66
GenBank mRNA D83044.1 - 6 - 7 1155 66
X98334.1 - 6 - 7 1101 66
Y13154.1 - 6 - 7 1102 66

View Full Product Details