The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031746.1 | NP_113934.1 | 11 - 12 | 1664 | 60 |
XM_006246947.2 | XP_006247009.1 | 11 - 12 | 1656 | 60 | |
GenBank mRNA | AB001321.1 | - | 11 - 12 | 1667 | 60 |
AF058714.1 | - | 11 - 12 | 1664 | 60 | |
U51153.1 | - | 11 - 12 | 1656 | 60 | |
X59677.1 | - | - | 391 | 60 |