Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcx
Entrez Gene ID
84394
Gene Name
doublecortin
Gene Aliases
-
Chromosome Location
Chr.X: 115098675 - 115175517 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053379.3 NP_445831.3 2 - 3 433 110
XM_006257382.2 XP_006257444.1 2 - 3 558 110
XM_006257383.2 XP_006257445.1 2 - 3 651 110
XM_006257384.2 XP_006257446.1 2 - 3 650 110
XM_006257385.2 XP_006257447.1 2 - 3 651 110
XM_006257386.2 XP_006257448.1 2 - 3 651 110
XM_008773448.1 XP_008771670.1 2 - 3 1120 110
GenBank mRNA AF155959.2 - 1 - 2 367 110

View Full Product Details